Learn More
Grainger 10", Men's, Overboots, Plain Toe Type, PVC Upper Material, Yellow, Fits Shoe Size 6-1/2 to 8
Catalog No. NC1667405
Encompass_Preferred
Workbrutes PVC overshoes are designed with thick, deep cleats for optimum traction and long service life. The injection-molded design features 46% more outsole material than comparable overshoes. The Frigiflex PVC compound contains special additives for high stretch and flexibility in cold temperatures. Use in food processing, construction, general industry, transportation, and hazardous waste cleanup. Overshoes offer resistance to fats, certain acids, hydrocarbons, and caustics. Not recommended for use with ketones, aldehydes, and many solvents.
This item is not returnable. View return policy
Catalog No. NC1667405 Supplier Grainger Supplier No. 9AHP7
Workbrutes PVC overshoes are designed with thick, deep cleats for optimum traction and long service life. The injection-molded design features 46% more outsole material than comparable overshoes. The Frigiflex PVC compound contains special additives for high stretch and flexibility in cold temperatures. Use in food processing, construction, general industry, transportation, and hazardous waste cleanup. Overshoes offer resistance to fats, certain acids, hydrocarbons, and caustics. Not recommended for use with ketones, aldehydes, and many solvents.