Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Hexylphenothiazine 98.0+%, TCI America™
SDP

Catalog No. H16085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16085G 5 g
H16081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16085G Supplier TCI America Supplier No. H16085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73025-93-1
Molecular Formula C18H21NS
Molecular Weight (g/mol) 283.433
InChI Key VRJVOTTZESKWPD-UHFFFAOYSA-N
PubChem CID 18972631
IUPAC Name 10-hexylphenothiazine
SMILES CCCCCCN1C2=CC=CC=C2SC3=CC=CC=C31

Specifications

Color Yellow
Quantity 5 g
Formula Weight 283.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-Hexylphenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.