Learn More
Bellco Glass, Inc. 10 Hole Large Flange Headplate for Omni Vessels
SDP
Catalog No. 501944898
Encompass_Preferred
200mm large flange headplate 10-port6@ 1/4" OD4@ 1/2" OD1 center for drive motor1964-90115 1/4" compression fitting1964-90120 1/2" compression fitting1964-90125 5/16" compression fitting (thermowell)Headplates are used with the large flange vessels to seal and to also provide ports for access to the interior of the vessel. The 18-port headplate is most used since more components can be fitted into its ports. The impeller is always positioned in the center drive port and a variety of components i.e. probe (1/2"), harvesting tubes (1/4" or 1/2"), thermowells (5/16") can be added as needed. Headplates and all stainless components of the bioreactor systems are electropolished to seal the surface against bacterial contamination that might occur with a rough surface. MADE IN THE USA. O-ring replacements: a554-30 6 eacha554-42 4 eacha554-13 1 each
Catalog No. 50-194-4898 Supplier Bellco Glass, Inc. Supplier No. 196488505
200mm large flange headplate 10-port6@ 1/4" OD4@ 1/2" OD1 center for drive motor1964-90115 1/4" compression fitting1964-90120 1/2" compression fitting1964-90125 5/16" compression fitting (thermowell)Headplates are used with the large flange vessels to seal and to also provide ports for access to the interior of the vessel. The 18-port headplate is most used since more components can be fitted into its ports. The impeller is always positioned in the center drive port and a variety of components i.e. probe (1/2"), harvesting tubes (1/4" or 1/2"), thermowells (5/16") can be added as needed. Headplates and all stainless components of the bioreactor systems are electropolished to seal the surface against bacterial contamination that might occur with a rough surface. MADE IN THE USA. O-ring replacements: a554-30 6 eacha554-42 4 eacha554-13 1 each