Learn More
MSC Irwin Step Drill Bit, 10 Hole Sizes, 1/4-1-3/8, 1 ea
Catalog No. NC1908039
Encompass
This high-speed steel step drill bit features 10 hole sizes ranging from 1/4 inch to 1-3/8 inch, designed for general-purpose drilling in materials like wood, aluminum, brass, mild steel, copper, laminate, and plexiglass.
  • Made from high-speed steel
  • Provides 10 distinct hole sizes
  • Features a hex shank for secure grip
  • Drills hole diameters from 1/4 in to 1-3/8 in
  • Step length of 1/8 in
  • Bright/uncoated finish for general purpose use
Catalog No. NC1908039 Supplier MSC Supplier No. 06560668
This high-speed steel step drill bit features 10 hole sizes ranging from 1/4 inch to 1-3/8 inch, designed for general-purpose drilling in materials like wood, aluminum, brass, mild steel, copper, laminate, and plexiglass.
  • Made from high-speed steel
  • Provides 10 distinct hole sizes
  • Features a hex shank for secure grip
  • Drills hole diameters from 1/4 in to 1-3/8 in
  • Step length of 1/8 in
  • Bright/uncoated finish for general purpose use