Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-Hydroxy-12(Z)-octadecenoic acid | 34932-12-2 | 99.8% | 298.46 | 1 MG
SDP

Catalog No. 5000391797
Encompass_Preferred

10-Hydroxy-12(Z)-octadecenoic acid is an ester product intended for research applications. This compound exhibits a molecular weight of 298.46 and a high purity of 99.82%. Researchers should note that it may cause skin irritation, allergic skin reactions, and serious eye irritation.

  • Ester product.
  • Suitable for research applications.
  • Molecular weight of 298.46.
  • Purity of 99.82%.
  • Identified by CAS number 34932-12-2.

Catalog No. 50-003-91797 Supplier Medchemexpress LLC Supplier No. HY1657721MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10-Hydroxy-12(Z)-octadecenoic acid is an ester product intended for research applications. This compound exhibits a molecular weight of 298.46 and a high purity of 99.82%. Researchers should note that it may cause skin irritation, allergic skin reactions, and serious eye irritation.

  • Ester product.
  • Suitable for research applications.
  • Molecular weight of 298.46.
  • Purity of 99.82%.
  • Identified by CAS number 34932-12-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.