Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-Hydroxy-12(Z)-octadecenoic acid | 34932-12-2 | 99.8% | 298.46 | 50 MG
SDP

Catalog No. 5000391809
Encompass_Preferred

10-Hydroxy-12(Z)-octadecenoic acid is presented as an ester product, appearing as a solid with a white to off-white color. It has a molecular weight of 298.46 and a chemical formula of C18H34O3. The CAS number for this compound is 34932-12-2. It is specified for research use only and not sold to patients. The purity of the product is 99.82%. For shipping, it is handled at room temperature within the continental US, with variations possible elsewhere. Storage recommendations include keeping the powder at -20°C for 3 years, and in solvent at -80°C for 6 months or -20°C for 1 month.

  • Purity: 99.82%.
  • Appearance: Solid, white to off-white.
  • Research use: For research use only; not sold to patients.

Catalog No. 50-003-91809 Supplier Medchemexpress LLC Supplier No. HY16577250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10-Hydroxy-12(Z)-octadecenoic acid is presented as an ester product, appearing as a solid with a white to off-white color. It has a molecular weight of 298.46 and a chemical formula of C18H34O3. The CAS number for this compound is 34932-12-2. It is specified for research use only and not sold to patients. The purity of the product is 99.82%. For shipping, it is handled at room temperature within the continental US, with variations possible elsewhere. Storage recommendations include keeping the powder at -20°C for 3 years, and in solvent at -80°C for 6 months or -20°C for 1 month.

  • Purity: 99.82%.
  • Appearance: Solid, white to off-white.
  • Research use: For research use only; not sold to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.