Learn More
Cayman Chemical 10-hydroxy DecnoIc AcId 10g
SDP
Catalog No. 502992132
Encompass_Preferred
A hydroxy fatty acid and a major component of worker bee mandibular blend pheromones; reduces LPS-induced nitric oxide production in RAW 264.7 cells at 5 mM
Catalog No. 50-299-2132 Supplier Cayman Chemical Supplier No. 3045810 G
A hydroxy fatty acid and a major component of worker bee mandibular blend pheromones; reduces LPS-induced nitric oxide production in RAW 264.7 cells at 5 mM