Learn More
Cayman Chemical 10-hydroxy WarfarIn 1mg
SDP
Catalog No. 502948093
Encompass_Preferred
A metabolite of (R)-warfarin; an inhibitor of CYP2C9 (IC50 = 1.6 µM)
Catalog No. 50-294-8093 Supplier Cayman Chemical Supplier No. 339291 MG
A metabolite of (R)-warfarin; an inhibitor of CYP2C9 (IC50 = 1.6 µM)