Learn More
Cayman Chemical 10-hydroxy WarfarIn 500ug
SDP
Catalog No. 502946781
Encompass_Preferred
A metabolite of (R)-warfarin; an inhibitor of CYP2C9 (IC50 = 1.6 µM)
Catalog No. 50-294-6781 Supplier Cayman Chemical Supplier No. 33929500 UG
A metabolite of (R)-warfarin; an inhibitor of CYP2C9 (IC50 = 1.6 µM)