Learn More
Selleck Chemical LLC 10-Hydroxydecanoic acid S5370-25mg
SDP
Catalog No. 503109486
Encompass_Preferred
10-Hydroxydecanoic acid a minor constituent of royal jelly is a saturated fatty acid that has cytotoxic effects
Catalog No. 50-310-9486 Supplier Selleck Chemical LLC Supplier No. S537025MG
10-Hydroxydecanoic acid a minor constituent of royal jelly is a saturated fatty acid that has cytotoxic effects