Learn More
US Plastic Corporation Utility Funnel, 10-inch Top Diameter, Natural 
SDP
Catalog No. NC9002177
Encompass
Extra Large Funnel With Large Throat, ideal for use with powders, granular material, and viscous liquids, flexible, non-corroding, non-contaminating low density polyethylene,Large Throat Funnel 10 in. Top Dia, 1- 3/4 in. Spout OD Dia.
Catalog No. NC9002177 Supplier US Plastic Corporation Supplier No. 78109
Extra Large Funnel With Large Throat, ideal for use with powders, granular material, and viscous liquids, flexible, non-corroding, non-contaminating low density polyethylene,Large Throat Funnel 10 in. Top Dia, 1- 3/4 in. Spout OD Dia.