Learn More
Buehler MicroCloth, PSA, 10 in
Catalog No. NC1569517
Encompass
This item has a minimum qty of 4 per supplier requirements.
MicroCloth is a soft, versatile, long napped, synthetic rayon cloth. It is an excellent general use cloth for final polishing stages for a wide variety of materials. Typical applications include but are not limited to ferrous and non-ferrous metals, ceramics, composites, PCBs, cast irons, cermets, plastics and electronics.
This item is not returnable. View return policy
Catalog No. NC1569517 Supplier Buehler Supplier No. 407220
This item has a minimum qty of 4 per supplier requirements.
MicroCloth is a soft, versatile, long napped, synthetic rayon cloth. It is an excellent general use cloth for final polishing stages for a wide variety of materials. Typical applications include but are not limited to ferrous and non-ferrous metals, ceramics, composites, PCBs, cast irons, cermets, plastics and electronics.