Learn More
Electron Microscopy Sciences 10 mcL/1 mcL Loop Natural Individually Wrapped
SDP
Catalog No. 5030205
Encompass_Preferred
Disposable incubation loops and needles eliminating the need for flaming metal loops and exposure to airborne pathogens. Precision molded from biological inert polymers. Irradiation sterilized. Disposable incubation loops and needles are Irradiation sterilized.
Catalog No. 50-302-05 Supplier Electron Microscopy Sciences Supplier No. 6244702
Disposable incubation loops and needles eliminating the need for flaming metal loops and exposure to airborne pathogens. Precision molded from biological inert polymers. Irradiation sterilized. Disposable incubation loops and needles are Irradiation sterilized.