Learn More
Electron Microscopy Sciences 10 mcL/1 mcL Loop/Needle Natural
SDP
Catalog No. 5030202
Encompass_Preferred
Disposable incubation loops and needles eliminating the need for flaming metal loops and exposure to airborne pathogens. Precision molded from biological inert polymers. Irradiation sterilized.
Catalog No. 50-302-02 Supplier Electron Microscopy Sciences Supplier No. 6244703
Disposable incubation loops and needles eliminating the need for flaming metal loops and exposure to airborne pathogens. Precision molded from biological inert polymers. Irradiation sterilized.