Learn More
Analytical Sales & Services 10µL GC Syringe for LEAP/Gerstel CTC PAL GC Autosamplers
Catalog No. 5000804814
Encompass
10µL GC Syringe 701N for LEAP/Gerstel CTC PAL GC Autosamplers. 26s Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH
Catalog No. 50-008-04814 Supplier Analytical Sales & Services Supplier No. 42032054
Add to Cart
Add to Cart
10µL GC Syringe 701N for LEAP/Gerstel CTC PAL GC Autosamplers. 26s Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH