Learn More
Chemglass Life Sciences 10µl Inoculating Loop, Blue, Sterile, Individual Pack
SDP
Catalog No. 5000083123
Encompass_Preferred
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification. Available in three styles; needle, 1ul loop and 10ul loop. Vacuum-gas plasma surface treated. Packaged in peel-to-open paper plastic wrappers. Product is sterile (gamma irradiated).
Catalog No. 50-000-83123 Supplier Chemglass Life Sciences Supplier No. CLS2458012
Loops and needles are made from high quality polypropylene and are flexible so stem can be bent to access small volume tubes and dishes. Color coded for easy identification. Available in three styles; needle, 1ul loop and 10ul loop. Vacuum-gas plasma surface treated. Packaged in peel-to-open paper plastic wrappers. Product is sterile (gamma irradiated).