Learn More
CELLTREAT Scientific Products 10µL Low Retention Pipette Tips, Racked, Non-sterile, Case Quantity 960
SDP
Catalog No. 5000670024
Encompass_Preferred
CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing processes results in superior low retention
Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting
Multiple Pipette Tip sizes are available, including extended length tips (10μL and 1000μL) to allow better access to the bottom of test tubes, reagent bottles, flasks and other vessels
Pipette Tip racks feature hinged lids that are designed for asy one-handed opening, making it easier to access repeatedly
Pipette Tips are universal fit and compatible with major pipettes
Fine tip for improved accuracy and precision
Molded volume reference markings offer a quick double check opportunity
Catalog No. 50-006-70024 Supplier CELLTREAT Scientific Products Supplier No. 229032NS
CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing processes results in superior low retention
Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting
Multiple Pipette Tip sizes are available, including extended length tips (10μL and 1000μL) to allow better access to the bottom of test tubes, reagent bottles, flasks and other vessels
Pipette Tip racks feature hinged lids that are designed for asy one-handed opening, making it easier to access repeatedly
Pipette Tips are universal fit and compatible with major pipettes
Fine tip for improved accuracy and precision
Molded volume reference markings offer a quick double check opportunity