Learn More
Analytical Sales & Services 10µL PEEK Sample Loop
Catalog No. 5000804965
Encompass
10µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH
Catalog No. 50-008-04965 Supplier Analytical Sales & Services Supplier No. 490550214
10µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH