Learn More
Analytical Sales & Services 10µL Syringe Model 701, Cemented Needle
Catalog No. 5000804299
Encompass
10µL Syringe Model 701, Cemented Needle, Point Style #3,, 2 in., 22s Gauge\n\nQty/Pkg: EACH
Catalog No. 50-008-04299 Supplier Analytical Sales & Services Supplier No. 4803654
10µL Syringe Model 701, Cemented Needle, Point Style #3,, 2 in., 22s Gauge\n\nQty/Pkg: EACH