Learn More
GSC Go Science Crazy 10 Level Conductivity Tester
SCI_ED
Catalog No. S28110 Shop All GSC Products
Click to view available options
Description:
10-level
Catalog No. Description
S28110 10-level
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S28110 Supplier GSC Go Science Crazy Supplier No. CNDTEST
Not a toy. Product intended for ages 13 and up.
Tool for quickly and easily determining the conductivity of various solutions. The 10 Level Conductivity Indicator can be used for quick comparisons of conductivity in solutions such as tap water, vinegar, baking soda solution, or varying concentrations of salt water.
  • Quickly compare conductivity
  • Activated by push and hold switch
  • Multicolored bar LED lights to display results and 3 in. brass prongs
Specifications
Description 10-level
Product Type Conductivity tester
Includes Multicolored bar LEDs to display results, 3 in. brass prongs
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov