Learn More
Eagle Stainless Container Eagle Stainless, 10 Liter Container with Lid, Gasket and Lock Lever Ring, 316L Stainless Steel. (CTL Series)
SDP
Catalog No. NC1590759
Encompass_Preferred
10 Liter container with lid, platinum cured silicone gasket and lock lever ring. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.
Catalog No. NC1590759 Supplier Eagle Stainless Container Supplier No. CTL24316LJ
10 Liter container with lid, platinum cured silicone gasket and lock lever ring. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.