Learn More
Analytical Sales & Services 10 Liter Waste Container, B83 Carboy
Catalog No. 5000805202
Encompass
10 Liter Waste Container, B83 Carboy\n\n• HDPE\n• B83 Threads\n• Round with Handles\n• Bottle Outer Diameter: 249mm\n• Bottle Height: 389mm\n• Molded Graduations\n• Includes White PP B83 Screw Cap (generic, not Canary-Safe)\n\nQty/Pkg: EACH
Catalog No. 50-008-05202 Supplier Analytical Sales & Services Supplier No. CL83010
10 Liter Waste Container, B83 Carboy\n\n• HDPE\n• B83 Threads\n• Round with Handles\n• Bottle Outer Diameter: 249mm\n• Bottle Height: 389mm\n• Molded Graduations\n• Includes White PP B83 Screw Cap (generic, not Canary-Safe)\n\nQty/Pkg: EACH