Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-methoxycamptothecin | 19685-10-0 | 98.1% | 378.38 g·mol⁻¹ | C21H18N2O5 | 10 MG
SDP

Catalog No. 5000295945
Encompass_Preferred

10-Methoxycamptothecin is a natural bioactive derivative of camptothecin used as an antitumor research reagent. It functions as a small-molecule topoisomerase I inhibitor and has shown higher cytotoxicity than 10-hydroxycamptothecin in cell-line studies, making it suitable for cytotoxicity assays and as an ADC payload in research.

  • CAS number: 19685-10-0.
  • Molecular formula: C21H18N2O5.
  • Molecular weight: 378.38 g·mol⁻¹.
  • Appearance: white to yellow solid.
  • Purity: 98.06%.
  • Storage: protect from light; store at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Typical applications: antitumor studies, cytotoxicity assays, ADC payload development.

Catalog No. 50-002-95945 Supplier Medchemexpress LLC Supplier No. HYN044610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

10-Methoxycamptothecin is a natural bioactive derivative of camptothecin used as an antitumor research reagent. It functions as a small-molecule topoisomerase I inhibitor and has shown higher cytotoxicity than 10-hydroxycamptothecin in cell-line studies, making it suitable for cytotoxicity assays and as an ADC payload in research.

  • CAS number: 19685-10-0.
  • Molecular formula: C21H18N2O5.
  • Molecular weight: 378.38 g·mol⁻¹.
  • Appearance: white to yellow solid.
  • Purity: 98.06%.
  • Storage: protect from light; store at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Typical applications: antitumor studies, cytotoxicity assays, ADC payload development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.