Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl)acridinium fluorosulfonate | 149300-54-9 | MFCD01320487 | >=90.0% | 413.42 g/mol | C21H16FNO5S | 5 MG
SDP
Catalog No. 5000281116
Encompass_Preferred
10-Methyl-9-(phenoxycarbonyl)acridinium fluorosulfonate is an acridinium ester-derived chemiluminescent probe that produces light under neutral conditions and is used for sensitive measurement of hydrogen peroxide in laboratory research settings. For research use only; not intended for human use.
  • Chemiluminescent probe for hydrogen peroxide measurement.
  • Produces chemiluminescence under neutral conditions.
  • High sensitivity suitable for biochemical assays.
  • Reported purity >=90.0% (HPLC).
  • Molecular formula C21H16FNO5S; molecular weight 413.42 g/mol.
  • Available in small research pack sizes, suitable for method development.
Catalog No. 50-002-81116 Supplier Medchemexpress LLC Supplier No. HYW4021485MG
10-Methyl-9-(phenoxycarbonyl)acridinium fluorosulfonate is an acridinium ester-derived chemiluminescent probe that produces light under neutral conditions and is used for sensitive measurement of hydrogen peroxide in laboratory research settings. For research use only; not intended for human use.
  • Chemiluminescent probe for hydrogen peroxide measurement.
  • Produces chemiluminescence under neutral conditions.
  • High sensitivity suitable for biochemical assays.
  • Reported purity >=90.0% (HPLC).
  • Molecular formula C21H16FNO5S; molecular weight 413.42 g/mol.
  • Available in small research pack sizes, suitable for method development.