Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) | 149300-54-9 | MFCD01320487 | 99.8% | 413.42 | C21H16FNO5S | 10 MG
SDP
Catalog No. 5000281055
Encompass_Preferred
10-Methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) is an acridinium ester reagent that produces chemiluminescence under neutral conditions and is used for measurement of hydrogen peroxide. Supplied as a light-yellow solid for research use; solutions are unstable and should be prepared fresh.
  • Chemiluminescent probe for hydrogen peroxide detection.
  • Phenyl ester acridinium structure for sensitive signal generation.
  • High purity suitable for analytical and research applications.
  • Light yellow solid for convenient solid-state handling.
  • Unstable in solution; prepare fresh solutions before use.
  • Recommended storage sealed at 4°C, protected from moisture and light.
Catalog No. 50-002-81055 Supplier Medchemexpress LLC Supplier No. HYW40214810MG
10-Methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) is an acridinium ester reagent that produces chemiluminescence under neutral conditions and is used for measurement of hydrogen peroxide. Supplied as a light-yellow solid for research use; solutions are unstable and should be prepared fresh.
  • Chemiluminescent probe for hydrogen peroxide detection.
  • Phenyl ester acridinium structure for sensitive signal generation.
  • High purity suitable for analytical and research applications.
  • Light yellow solid for convenient solid-state handling.
  • Unstable in solution; prepare fresh solutions before use.
  • Recommended storage sealed at 4°C, protected from moisture and light.