Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl)acridinium fluorosulfonate | 149300-54-9 | MFCD01320487 | 99.8% | 413.42 | C21H16FNO5S | 25 MG
SDP
Catalog No. 5000281019
Encompass_Preferred
10-Methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) is an acridinium-based chemiluminescent probe used for detection and measurement of hydrogen peroxide and reactive oxygen species in research applications. It is supplied as a light yellow to yellow solid with high purity and should be stored sealed at 4°C away from moisture and light.
  • Chemiluminescent probe for hydrogen peroxide and ROS detection.
  • High purity suitable for research applications.
  • Light yellow to yellow solid with defined physical appearance.
  • Molecular weight 413.42 and formula C21H16FNO5S for compound identification.
  • Available in small pack sizes for assay development and optimization.
Catalog No. 50-002-81019 Supplier Medchemexpress LLC Supplier No. HYW40214825MG
10-Methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) is an acridinium-based chemiluminescent probe used for detection and measurement of hydrogen peroxide and reactive oxygen species in research applications. It is supplied as a light yellow to yellow solid with high purity and should be stored sealed at 4°C away from moisture and light.
  • Chemiluminescent probe for hydrogen peroxide and ROS detection.
  • High purity suitable for research applications.
  • Light yellow to yellow solid with defined physical appearance.
  • Molecular weight 413.42 and formula C21H16FNO5S for compound identification.
  • Available in small pack sizes for assay development and optimization.