Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) | 149300-54-9 | MFCD01320487 | 99.8% | 413.42 | C21H16FNO5S | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW40214850MG
Encompass_Preferred
10-Methyl-9-(phenoxycarbonyl)acridinium (fluorosulfonate) is an acridinium-based chemiluminescent reagent used in research to detect and quantify hydrogen peroxide. As a phenyl ester of acridinium, it produces light under neutral conditions and is provided at high purity for reliable analytical and assay performance.
  • Produces chemiluminescence under neutral conditions.
  • Sensitive probe for hydrogen peroxide detection.
  • High purity for reproducible analytical results.
  • Available in small pack sizes for laboratory use.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-81335