Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate | 161006-14-0 | MFCD01320487 | 99.4% | 463.43 | C22H16F3NO5S | 5 MG
SDP
Catalog No. 5000281079
Encompass_Preferred
10-Methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate is an acridinium-based chemiluminescent reagent used in chemiluminescent assays for sensitive detection of enzymes, antigens, antibodies, and hormones. It converts to a fluorescent acridone upon oxidation and is supplied in small quantities for research and assay development.
Catalog No. 50-002-81079 Supplier Medchemexpress LLC Supplier No. HYW4151085MG
10-Methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate is an acridinium-based chemiluminescent reagent used in chemiluminescent assays for sensitive detection of enzymes, antigens, antibodies, and hormones. It converts to a fluorescent acridone upon oxidation and is supplied in small quantities for research and assay development.