Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate | 161006-14-0 | MFCD32173632 | 99.4% | 463.43 g/mol | C22H16F3NO5S | 10 MG
SDP
Catalog No. 5000281039
Encompass_Preferred
10-Methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate is an acridinium ester supplied as a trifluoromethanesulfonate salt that produces fluorescent 10-methyl-9-acridone upon oxidation. It is used as a chemiluminescent probe for sensitive detection in biochemical and environmental analyses.
  • Produces fluorescent acridone upon oxidation for signal generation.
  • Suitable for chemiluminescent and enzyme-linked assay formats.
  • Compatible with antigen, antibody, and hormone immunoassays.
  • Enables detection of oxidants in environmental and biological samples.
  • Provided as a stable, crystalline reagent for research use.
Catalog No. 50-002-81039 Supplier Medchemexpress LLC Supplier No. HYW41510810MG
10-Methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate is an acridinium ester supplied as a trifluoromethanesulfonate salt that produces fluorescent 10-methyl-9-acridone upon oxidation. It is used as a chemiluminescent probe for sensitive detection in biochemical and environmental analyses.
  • Produces fluorescent acridone upon oxidation for signal generation.
  • Suitable for chemiluminescent and enzyme-linked assay formats.
  • Compatible with antigen, antibody, and hormone immunoassays.
  • Enables detection of oxidants in environmental and biological samples.
  • Provided as a stable, crystalline reagent for research use.