Learn More
Medchemexpress LLC 10-methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate | 161006-14-0 | MFCD32173632 | 99.4% | 463.43 g/mol | C22H16F3NO5S | 25 MG

Supplier: Medchemexpress LLC HYW41510825MG
10-Methyl-9-(phenoxycarbonyl) acridinium trifluoromethanesulfonate is an acridinium ester chemiluminescent probe that produces fluorescent 10-methyl-9-acridone upon oxidation. It is designed for sensitive detection workflows, including chemiluminescent assays and immunoassays, and for measuring oxidants in biological and environmental samples.
- High purity: 99.4%.
- Molecular weight 463.43 g/mol.
- Chemical formula C22H16F3NO5S.
- Suitable for chemiluminescent and immunoassays.
- Useful for oxidant detection in biological and environmental samples.
- Supplied as a 25 MG pack for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.