Learn More
Accuflow Systems Sterile Empty Vial (SEV), 10 mL, 20 mm clear glass with green cap - 25/pack
Catalog No. NC1741082
Encompass_Preferred
ALK Sterile Empty Vials (SEV) feature Type 1 glass USP borosilicate, and a latex free stopper. ALK SEVs are produced at a cGMP FDA Manufacturer and USP tested for sterility and endotoxins. Certificate of Analysis available for all production lots and vials are aseptically assembled in an ISO 5 (class 100) cleanroom.
Catalog No. NC1741082 Supplier Accuflow Systems Supplier No. SEV1020G
ALK Sterile Empty Vials (SEV) feature Type 1 glass USP borosilicate, and a latex free stopper. ALK SEVs are produced at a cGMP FDA Manufacturer and USP tested for sterility and endotoxins. Certificate of Analysis available for all production lots and vials are aseptically assembled in an ISO 5 (class 100) cleanroom.