Learn More
Perkin Elmer US LLC 10 mL BD Luer-Lok Disposable Syringe PK/100
Catalog No. 502113404
Encompass_Preferred
Clear Nylon Syringe Filters, 0. 22 µm, 4 mm (200/Pack) are ideal for aqueous (non-acidic) or organic sample prep and HPLC, GC or dissolution sample analysis. With its excellent flow characteristics, very low extractable levels and mechanical stability, Nylon offers the best combination of physical parameters to meet the most stringent analytical needs. The naturally hydrophilic, high protein binding and high dirt loading capacity of Nylon are natural advantages. All our syringe filters are HPLC certified. Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis
Catalog No. 50-211-3404 Supplier Perkin Elmer US LLC Supplier No. 02542893
Clear Nylon Syringe Filters, 0. 22 µm, 4 mm (200/Pack) are ideal for aqueous (non-acidic) or organic sample prep and HPLC, GC or dissolution sample analysis. With its excellent flow characteristics, very low extractable levels and mechanical stability, Nylon offers the best combination of physical parameters to meet the most stringent analytical needs. The naturally hydrophilic, high protein binding and high dirt loading capacity of Nylon are natural advantages. All our syringe filters are HPLC certified. Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis