Learn More
Ace Glass, Inc. 10mL buret, cs/1, 2122A-10; PYREX brand, with Accu-RedTM markings. Suitable for use in schools and institutional laboratories not requiring the accuracy of Class A models.
SDP
Catalog No. NC9954632
Encompass
10mL buret, cs/1, 2122A-10; PYREX brand, with Accu-RedTM markings. Suitable for use in schools and institutional laboratories not requiring the accuracy of Class A models.
Catalog No. NC9954632 Supplier Ace Glass, Inc. Supplier No. 576560
10mL buret, cs/1, 2122A-10; PYREX brand, with Accu-RedTM markings. Suitable for use in schools and institutional laboratories not requiring the accuracy of Class A models.