Learn More
Metrohm USA 10 ML DOSING UNIT
SDP
Supplier:  Metrohm USA 63032210
Encompass
Dosing unit with integrated data chip with 10 mL glass cylinder and light protection, mountable on reagent bottle with ISO/DIN GL45 thread. FEP tubing connection, anti-diffusion buret tip.
Catalog No. NC0710632