Learn More
Andwin Scientific 10 ML PP STORAGE VIAL WITH SCR
SDP
Catalog No. 50819837
Encompass_Preferred
The 10 ml conical vial features a self-standing design, graduations that allow precise sample measurement, and a large, etched writing surfacethat permits easy identification. This product also offers secure storage for the sample with the attached HDPE screw cap with a sealing ring. It comes in a pack of 2500 vials.
Catalog No. 50-819-837 Supplier Andwin Scientific Supplier No. 15101000
The 10 ml conical vial features a self-standing design, graduations that allow precise sample measurement, and a large, etched writing surfacethat permits easy identification. This product also offers secure storage for the sample with the attached HDPE screw cap with a sealing ring. It comes in a pack of 2500 vials.