Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotage 10 ML REACTOR VIAL W/PTFE FRIT
SDP

Catalog No. NC1867139
Encompass

  • Capacity: 10 mL
  • Frit material: PTFE
  • Vial material: Microwave-safe glass
  • Pressure resistance: >30 bar
  • Max pressure (specific conditions): 20 bar (for 10-20 mL vials when >250 °C)
  • Pack quantity: 50 vials
  • Compatibility: Biotage Initiator+; Initiator microwave synthesizers
  • Primary application: Microwave-assisted synthesis
  • Specific applications: Peptide synthesis; Organic synthesis
  • Material feature: Contaminant-free glass
  • Design feature: Integrated PTFE frit
  • Durability: Engineered for high durability
  • Mixing method: Supports magnetic stirring
  • Scalability range: 0.2–20 mL (as part of the series)
  • Scale suitability: Multi-gram scale (for 10-20 mL vials)
  • Usage type: Disposable

Catalog No. NC1867139 Supplier Biotage Supplier No. 356289
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Capacity: 10 mL
  • Frit material: PTFE
  • Vial material: Microwave-safe glass
  • Pressure resistance: >30 bar
  • Max pressure (specific conditions): 20 bar (for 10-20 mL vials when >250 °C)
  • Pack quantity: 50 vials
  • Compatibility: Biotage Initiator+; Initiator microwave synthesizers
  • Primary application: Microwave-assisted synthesis
  • Specific applications: Peptide synthesis; Organic synthesis
  • Material feature: Contaminant-free glass
  • Design feature: Integrated PTFE frit
  • Durability: Engineered for high durability
  • Mixing method: Supports magnetic stirring
  • Scalability range: 0.2–20 mL (as part of the series)
  • Scale suitability: Multi-gram scale (for 10-20 mL vials)
  • Usage type: Disposable

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.