Learn More
EXEL Medical Products 10 mL Syringe
Catalog No. 01001680 Shop All Air-Tite Products
Change view
Click to view available options
Connection Type:
Luer Lock
Luer Slip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
01-001-680 Luer Slip
01-001-679 Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
NA
Catalog No. 01-001-680 Supplier EXEL Medical Products Supplier No. ES1026266
Add to Cart
Add to Cart
NA
These syringes offer unsurpassed quality at a reasonable price. They are latex-free, sterile, non-pyrogenic, and non-toxic.
  • These syringes are packaged without needles
  • Designed with a positive plunger stop to prevent spillage when drawing and a smooth plunger motion
  • The crystal-clear barrel has easy-to-read, large, bold print
  • Two tips are available including the luer lock with cap and the luer slip with cap
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe
Disposable Single-use
Volume (Metric) 10 mL
Connection Type Luer Slip
Graduated Yes
Sterility Sterile
NA