Learn More
Vici Precision Sampling, Inc. 10 ML SYRINGE BEVEL NEEDLE
Catalog No. NC9519350
Encompass_Preferred
A2 gas syringe, 10ml, RN .029 in. x .012 in. x 2 in., bevel open end
Catalog No. NC9519350 Supplier Vici Precision Sampling, Inc. Supplier No. 050036
A2 gas syringe, 10ml, RN .029 in. x .012 in. x 2 in., bevel open end