Learn More
Medchemexpress LLC WM-1119 KAT6A inhibitor | 2055397-28-7 | 10 mM * 1 mL in DMSO
SDP
Supplier:  Medchemexpress LLC HY102058
Encompass_Preferred
WM-1119 is a highly potent and selective KAT6A inhibitor, with an IC50 of 0.25 μM for KAT6A in lymphoma cells, the binding KD values of WM-1119 with KAT6A, KAT5 and KAT7 are 2 nM, 2.2 μM, 0.5 μM , respectively.
Catalog No. NC1901423