Learn More
Qiagen Beverly Inc 10 MM DATP SOLUTION MIX
Catalog No. 50305947
Encompass
10 mM each dNTP (dATP, dCTP, dGTP, dTTP) at pH 7.5
Catalog No. 50-305-947 Supplier Qiagen Beverly Inc Supplier No. N205010L
10 mM each dNTP (dATP, dCTP, dGTP, dTTP) at pH 7.5