Learn More
OCEAN OPTICS INC RT Transmission Dip Probe Removeable Tips for Ocean Optics T300 | 10mm transmission tip
Catalog No. NC1789390
Encompass
Removable stainless steel tips for T300 dip probe. Each tip is equipped with a front surface mirror for redirecting the light from the illumination fiber in the T300 probe to the receiving fiber. The optical pathlength is therefore 2X the geometric distance to the mirror.
Catalog No. NC1789390 Supplier OCEAN OPTICS INC Supplier No. RT10MM
Removable stainless steel tips for T300 dip probe. Each tip is equipped with a front surface mirror for redirecting the light from the illumination fiber in the T300 probe to the receiving fiber. The optical pathlength is therefore 2X the geometric distance to the mirror.