Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 10% NBF in StatClick Vials
SDP

Catalog No. 21010345
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-345 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-345 Supplier StatLab Supplier No. NB0230P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Get your tissue into fixative as quickly and securely as possible with prefilled containers.

  • 10% NBF in 60 mL StatClick Vials - 64 vials/flat
  • 60 mL vial is filled with 30 mL of 10% NBF - Neutral Buffered Formalin
  • Prefill Size, Width x Height: 52 x 58 mm
  • StatClick containers produce an audible clicking sound when the leak-proof seal has been properly engaged assuring accurate closure and maintaining specimen integrity during transport
  • Containers are 95 kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 60 mL
Solution Type Formalin
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.