Learn More
Leica Biosystems Leica Biosystems 10% Neutral Buffered Formalin, 2.5 Gallon
Catalog No. NC2752763
Encompass
10% Neutral Buffered Formalin is a ready-to-use fixative formulated for routine tissue fixation in histology and pathology laboratories. This product is supplied in a 2.5 gallon cube container for bulk laboratory use.
  • Ready-to-use 10% neutral buffered formalin for tissue fixation.
  • 2.5 gallon cube container (≈9.46 L) for bulk use.
  • Suitable for routine histology and pathology applications.
  • Premeasured volume reduces preparation time and variability.
  • Packaged for storage and transport in laboratory environments.
Catalog No. NC2752763 Supplier Leica Biosystems Supplier No. 3800602
10% Neutral Buffered Formalin is a ready-to-use fixative formulated for routine tissue fixation in histology and pathology laboratories. This product is supplied in a 2.5 gallon cube container for bulk laboratory use.
  • Ready-to-use 10% neutral buffered formalin for tissue fixation.
  • 2.5 gallon cube container (≈9.46 L) for bulk use.
  • Suitable for routine histology and pathology applications.
  • Premeasured volume reduces preparation time and variability.
  • Packaged for storage and transport in laboratory environments.