Learn More
Leica Biosystems 10% Neutral Buffered Formalin Fixative, 4 gal
Catalog No. NC1614788
Encompass_Preferred
Ready-to-use 10% Neutral Buffered Formalin is a phosphate-buffered fixative supplied in a 4-gallon container for routine histology tissue fixation. It preserves tissue morphology and is compatible with standard staining and downstream processing.
  • Prepared 10% formalin solution for consistent fixation.
  • Phosphate buffered to maintain neutral pH and tissue integrity.
  • Supplied in a 4-gallon container for bulk laboratory use.
  • Suitable for routine histology fixation and processing workflows.
  • Safety data sheet and product flyer available for handling and compliance.
Catalog No. NC1614788 Supplier Leica Biosystems Supplier No. 3800598
Ready-to-use 10% Neutral Buffered Formalin is a phosphate-buffered fixative supplied in a 4-gallon container for routine histology tissue fixation. It preserves tissue morphology and is compatible with standard staining and downstream processing.
  • Prepared 10% formalin solution for consistent fixation.
  • Phosphate buffered to maintain neutral pH and tissue integrity.
  • Supplied in a 4-gallon container for bulk laboratory use.
  • Suitable for routine histology fixation and processing workflows.
  • Safety data sheet and product flyer available for handling and compliance.