Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 10% Neutral Buffered
SDP

Catalog No. 21402148
Change view
Click to view available options
Quantity:
7 mL
20 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-148 7 mL
21-402-147 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-148 Supplier StatLab Supplier No. NB0735/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Prefilled in StatClick containers that feature an audible clicking sound, assures accurate closure and maintaining specimen integrity during transport

  • StatClick containers are ready-to-use and feature a pH level of 7.0
  • Prefills are filled 50% with 10% NBF fixative
  • 20 mL containers
  • 64 prefills per case

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type 10% Neutral Buffered Formalin
Quantity 7 mL
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.