Learn More
Scigen 10% Neutral Buffered Formalin
Catalog No. 23730632 Shop All Scigen Inc Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
23-730-632 Non-sterile
1 options
This item is not returnable. View return policy
For In Vitro Laboratory Use Only
Catalog No. 23-730-632 Supplier Scigen Supplier No. 5071
Add to Cart
Add to Cart
This item is not returnable. View return policy
For In Vitro Laboratory Use Only
Scigen™ 10% Neutral Buffered Formalin (3.8 L/1 gal) is in stock and available to ship immediately.
  • It is recommended to treat formalin waste with Neutralex™, a government certified solution, resulting in drain safe waste. Neutralex™ provides complete neutralization in 15 minutes with no pH adjustments needed. After treatment, Neutralex™ is certified to produce a non-toxic solution that can be disposed into the sewer system without fear of environmental damage.
    • 4046 Neutralex™ Starter Kit
    • 4047 Neutralex™ Case of 16 Premeasured Packets
    • 4304 Neutra-Form™ Bulk Spill Neutralizing Agent
    • 4278 Neutra-Wipes™ For Small Formalin Spills
    • 4295 Neutra-Pads™ Neutralizing Grossing Pads
    • 4050 Neutralex™ Aldehyde Test Kit
Specifications
Product Type Buffered Formalin
Sterility Non-sterile