Learn More
Alkali Scientific 10% Neutral Buffered Formalin (NBF) Specimen Containers, Pre Filled, Dual Click Lids, Leak Resistant, 95 kPa Compliant, 96/Case, 40mL (1.34oz) 3 x 32 Carry Packs
SDP
Catalog No. 5000663028
Encompass_Preferred
These formalin filled specimen containers are designed to provide reliable leak resistance and secure, air tight sealing for the fixation and transport of tissue specimens in histology and pathology laboratories. Each container is prefilled halfway with 10% neutral buffered formalin (NBF), ensuring consistent fixation while reducing handling steps and exposure risk. Manufactured from translucent polypropylene (PP), the containers allow clear visibility of contents and are paired with durable polyethylene (PE) caps that create a dependable, tight seal. Select s feature a positive lock closure with audible confirmation to indicate the container is fully secured for safe transport. For efficient storage and distribution, containers are packaged in inner packs of 24 units, with four inner packs per case, providing a total of 96 containers per case.
Catalog No. 50-006-63028 Supplier Alkali Scientific Supplier No. NBF40
These formalin filled specimen containers are designed to provide reliable leak resistance and secure, air tight sealing for the fixation and transport of tissue specimens in histology and pathology laboratories. Each container is prefilled halfway with 10% neutral buffered formalin (NBF), ensuring consistent fixation while reducing handling steps and exposure risk. Manufactured from translucent polypropylene (PP), the containers allow clear visibility of contents and are paired with durable polyethylene (PE) caps that create a dependable, tight seal. Select s feature a positive lock closure with audible confirmation to indicate the container is fully secured for safe transport. For efficient storage and distribution, containers are packaged in inner packs of 24 units, with four inner packs per case, providing a total of 96 containers per case.