Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers

Catalog No. 22110757 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Formalin
Specimen Storage Containers
Packaging Type:
Carton
Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Type
22-110-757 Formalin Carton
22-045-400 Formalin Tray
9990910 Specimen Storage Containers Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-110-757 Supplier Epredia™ Supplier No. 51201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use these containers for the transportation and short-term storage of patient specimens.

Features:
  • Seal-Rite containers are engineered to maintain seal at 95KPA – the industry standard for proper sealing and transportation via air shipment
  • Double-click feature provides audible confirmation that the lid has been aligned, tightened properly and sealed
  • Arrow and lock icons align to visually signify the containers have been properly sealed
  • Embossed mL measuring scale identifies volume of liquid in the container
  • Knurled cap and base of the jar for better gripping
  • Safety. Security. Peace of Mind – Avoid the loss of precious tissue samples and exposure of staff to chemicals caused by leaking containers
A properly closed Epredia Seal-Rite container is leak-proof.
The containers have been tested using industry-standard International Safe Transit Association (ISTA) performance validation. Internal tests were performed as well with 0% leakage rate.

Specifications

Capacity 20 mL
Packaging Type Carton
Product Type Formalin
Quantity 192/cs
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.