Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers

Catalog No. 9990910 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Formalin
Specimen Storage Containers
Packaging Type:
Carton
Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Type
9990910 Specimen Storage Containers Tray
22-110-757 Formalin Carton
22-045-400 Formalin Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 9990910 Supplier Epredia™ Supplier No. 9990910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these containers for the transportation and short-term storage of patient specimens.

  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available

Specifications

Capacity 20 mL
Packaging Type Tray
Product Type Specimen Storage Containers
Quantity 180/cs
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.