Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific 10% Neutral Buffered Formalin
SDP

Catalog No. 22026212
Change view
Click to view available options
Quantity:
80 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-026-212 80 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-026-212 Supplier Azer Scientific Supplier No. PFNBF1.25G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1.25 gallon container half filled with 10% neutral buffered formalin. Features a handle for easy transport. Container made from high-density polyethylene. Secure screw top closure creates a moisture- and tamper-resistant seal

Used in histological applications for fixation and storage of tissue specimens.

Specifications

Quantity 80 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.